tert-Butyl 4-(2-((methylsulfonyl)oxy)ethyl)piperidine-1-carboxylate ≥97%

Supplier: Ambeed
Warning

Synonyms: tert-butyl4-[2-(methanesulphonyloxy)ethyl]piperidine-1-carboxylate

A320069-250MG A320069-5G A320069-1G A320069-25G
77612-848EA 46.35 USD
77612-848 77612-850 77683-951 77683-952
tert-Butyl 4-(2-((methylsulfonyl)oxy)ethyl)piperidine-1-carboxylate ≥97%
tert-Butyl 4-(2-((methylsulfonyl)oxy)ethyl)piperidine-1-carboxylate

Precaución: For research use only.

Formula: C₁₃H₂₅NO₅S
MW: 307.40 g/mol
MDL Number: MFCD11007804
CAS Number: 147699-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR