all cis-8,11,14-Eicosatrienoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Dihomo-gamma-linolenic Acid, Dihomo-γ-linolenic acid, (8Z,11Z,14Z)-Icosa-8,11,14-trienoic acid, all cis-8,11,14-Icosatrienoic Acid

A320023-5MG A320023-100MG A320023-50MG
77626-586EA 59.74 USD
77626-586 77879-656 77879-657
all cis-8,11,14-Eicosatrienoic acid ≥98%
all cis-8,11,14-Eicosatrienoic acid

Precaución: for Research Use Only

Formula: C₂₀H₃₄O₂
MW: 306.49 g/mol
MDL Number: MFCD00065721
CAS Number: 1783-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR