Alverine citrate salt ≥98%

Supplier: Ambeed
Warning

A319955-1G A319955-25G A319955-5G
77616-408EA 11.33 USD
77616-408 77616-410 77616-412
Alverine citrate salt ≥98%
Alverine citrate salt

Precaución: for Research Use Only

Formula: C₂₆H₃₅NO₇
MDL Number: MFCD00035086
CAS Number: 5560-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR