4-Chloro-6-methyl-7-azaindole ≥98%

Supplier: Ambeed
Warning

A313989-100MG A313989-1G A313989-250MG
77672-602EA 68 USD
77672-602 77672-604 77672-606
4-Chloro-6-methyl-7-azaindole ≥98%
4-Chloro-6-methyl-7-azaindole

Precaución: for Research Use Only

Formula: C₈H₇ClN₂
MW: 166.61 g/mol
MDL Number: MFCD09880146
CAS Number: 171879-99-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR