2-Bromo-3,6-difluoropyridine ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A319296-250MG A319296-10G A319296-1G A319296-5G
77683-948EA 66 USD
77683-948 77890-638 77890-639 77890-640
2-Bromo-3,6-difluoropyridine ≥98%, Ambeed.Inc
2-Bromo-3,6-difluoropyridine

Precaución: For research use only.

Formula: C₅H₂BrF₂N
MDL Number: MFCD27939127
CAS Number: 1382786-22-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR