2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Cetirizine

A313360-100MG A313360-250MG
77626-564EA 55.62 USD
77626-564 77879-637
2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)acetic acid ≥98%
2-(2-(4-((4-Chlorophenyl)(phenyl)methyl)piperazin-1-yl)ethoxy)acetic acid

Precaución: for Research Use Only

Formula: C₂₁H₂₅ClN₂O₃
MW: 388.89 g/mol
MDL Number: MFCD00800721
CAS Number: 83881-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR