Fmoc-(R)-3-amino-4-(4-fluorophenyl)butyric acid ≥98%

Supplier: Ambeed
Warning

A318705-100MG A318705-1G A318705-250MG
77900-525EA 30 USD
77900-525 77900-515 77901-113
Fmoc-(R)-3-amino-4-(4-fluorophenyl)butyric acid ≥98%
Fmoc-(R)-3-amino-4-(4-fluorophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₂₅H₂₂FNO₄
MDL Number: MFCD01860915
CAS Number: 331763-70-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR