2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulfonyl)benzoic acid 95+%

Supplier: Ambeed
Warning

A312918-25G A312918-5G
77683-925EA 61 USD
77683-925 77683-926
2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulfonyl)benzoic acid 95+%
2-Ethoxy-5-((4-ethylpiperazin-1-yl)sulfonyl)benzoic acid

Precaución: For research use only.

Formula: C₁₅H₂₂N₂O₅S
MW: 342.41 g/mol
MDL Number: MFCD16877564
CAS Number: 1245644-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR