1-Methylbenzoimidazol-5-amine ≥97%

Supplier: Ambeed
Warning

A318640-250MG A318640-5G
77685-910EA 22 USD
77685-910 77685-911
1-Methylbenzoimidazol-5-amine ≥97%
1-Methylbenzoimidazol-5-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MDL Number: MFCD03164349
CAS Number: 10394-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR