3,4,4'-Trichlorocarbanilide ≥98%

Supplier: Ambeed
Danger

A312675-25G A312675-100G
77616-364EA 7.21 USD
77616-364 77748-610
3,4,4'-Trichlorocarbanilide ≥98%
3,4,4'-Trichlorocarbanilide

Precaución: for Research Use Only

Formula: C₁₃H₉Cl₃N₂O
MDL Number: MFCD00013254
CAS Number: 101-20-2
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR