4-(2-Bromoethoxy)benzeneboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (4-(2-Bromoethoxy)phenyl)boronic acid

A312561-100MG A312561-250MG A312561-5G
77883-851EA 8 USD
77883-851 77883-852 77883-853
4-(2-Bromoethoxy)benzeneboronic acid ≥98%
4-(2-Bromoethoxy)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₀BBrO₃
MDL Number: MFCD09027232
CAS Number: 913836-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR