3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid ≥95%
Supplier: Ambeed
|
Warning
|
A312288-100MG
A312288-1G
77890-612EA
204
USD
77890-612
77890-613
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid ≥95%
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₉NO₂ |
MDL Number:
MFCD18826734 CAS Number: 406947-32-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...