4,4,5,5-Tetramethyl-2-(1-methylcyclopropyl)-1,3,2-dioxaborolane ≥98%, Ambeed.Inc

Supplier: Ambeed
Danger

A312276-1G A312276-250MG
77683-922EA 421 USD
77683-922 77683-923
4,4,5,5-Tetramethyl-2-(1-methylcyclopropyl)-1,3,2-dioxaborolane ≥98%, Ambeed.Inc
4,4,5,5-Tetramethyl-2-(1-methylcyclopropyl)-1,3,2-dioxaborolane

Precaución: For research use only.

Formula: C₁₀H₁₉BO₂
MW: 182.06 g/mol
MDL Number: MFCD23381517
CAS Number: 126689-04-1
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR