2-Fluoro-4-(methylsulfonyl)phenylboronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Fluoro-4-(methylsulphonyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A312249-100MG A312249-1G A312249-250MG
77885-829EA 94 USD
77885-829 77885-830 77885-831
2-Fluoro-4-(methylsulfonyl)phenylboronic acid pinacol ester ≥95%
2-Fluoro-4-(methylsulfonyl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₄S
MDL Number: MFCD18760181
CAS Number: 1384951-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR