Encorafenib ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-methyl (1-((4-(3-(5-chloro-2-fluoro-3-(methylsulphonamido)phenyl)-1-isopropyl-1H-pyrazol-4-yl)pyrimidin-2-yl)amino)propan-2-yl)carbamate, LGX818

A312190-10MG A312190-50MG
77612-792EA 166.86 USD
77612-792 77612-794
Encorafenib ≥98%
Encorafenib

Precaución: for Research Use Only

Formula: C₂₂H₂₇ClFN₇O₄S
MW: 540.01 g/mol
MDL Number: MFCD25976758
CAS Number: 1269440-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR