(2-Formyl-3-hydroxyphenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A2438729-250MG A2438729-1G
77877-817EA 189 USD
77877-817 77877-816
(2-Formyl-3-hydroxyphenyl)boronic acid ≥95%
(2-Formyl-3-hydroxyphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₇BO₄
CAS Number: 2892327-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR