Iberin ≥98%
Supplier: Ambeed
|
Danger
|
A243802-50MG
A243802-100MG
A243802-10MG
A243802-250MG
A243802-1G
77600-966EA
431.53
USD
77600-966
77667-990
77667-992
77879-429
77879-428
Iberin ≥98%
Iberin
Precaución: for Research Use Only
Formula:
C₅H₉NOS₂ |
MDL Number:
MFCD01076593 CAS Number: 505-44-2 UN: 2810 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...