(3-Methyl-2-pyridinyl)methanamine ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-(Aminomethyl)-3-methylpyridine

A311999-1G A311999-5G
77593-868EA 44.08 USD
77593-868 77746-598
(3-Methyl-2-pyridinyl)methanamine ≥98%
(3-Methyl-2-pyridinyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.17 g/mol
MDL Number: MFCD04116209
CAS Number: 153936-26-6
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR