6-Chloro-2-pyridineboronic acid ≥95%

Supplier: Ambeed
Warning

Synonyms: 6-Chloropyridine-2-boronic Acid

A311829-100MG A311829-250MG
77626-556EA 52.53 USD
77626-556 77626-558
6-Chloro-2-pyridineboronic acid ≥95%
6-Chloro-2-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD08063028
CAS Number: 652148-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR