6-Methoxybenzo[c][1,2]oxaborol-1(3H)-ol 95+%

Supplier: Ambeed
Warning

A243447-100MG A243447-1G A243447-250MG
77890-395EA 107 USD
77890-395 77890-396 77890-397
6-Methoxybenzo[c][1,2]oxaborol-1(3H)-ol 95+%
6-Methoxybenzo[c][1,2]oxaborol-1(3H)-ol

Precaución: for Research Use Only

Formula: C₈H₉BO₃
MDL Number: MFCD16658744
CAS Number: 947163-26-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR