Sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate hydrate ≥98%

Supplier: Ambeed
Warning

A311515-5G A311515-10G A311515-1G
77626-552EA 585.04 USD
77626-552 77877-993 77877-994
Sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate hydrate ≥98%
Sodium 2,9-dimethyl-4,7-diphenyl-1,10-phenanthroline-3,8-disulfonate hydrate

Precaución: for Research Use Only

Formula: C₂₆H₂₀N₂Na₂O₇S₂
MDL Number: MFCD00149974
CAS Number: 1257642-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR