2-(1-Benzylpiperidin-4-yl)acetic acid ≥95%

Supplier: Ambeed
Warning

A311472-1G A311472-250MG A311472-5G A311472-100MG
77616-358EA 116.68 USD
77616-358 77616-360 77746-597 77746-596
2-(1-Benzylpiperidin-4-yl)acetic acid ≥95%
2-(1-Benzylpiperidin-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₂
MDL Number: MFCD13250102
CAS Number: 130927-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR