2-(2-Pyridyl)-2-propylamine Dihydrochloride ≥98%

Supplier: Ambeed
Warning

A311143-250MG A311143-1G
77685-887EA 40 USD
77685-887 77901-943
2-(2-Pyridyl)-2-propylamine Dihydrochloride ≥98%
2-(2-Pyridyl)-2-propylamine Dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄Cl₂N₂
MDL Number: MFCD18070980
CAS Number: 1256633-17-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR