5-Hydroxy-2-(4-hydroxyphenyl)-6-methoxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Homoplantaginin

A310971-10MG A310971-1MG A310971-5MG
77612-782EA 107.12 USD
77612-782 77612-784 77612-786
5-Hydroxy-2-(4-hydroxyphenyl)-6-methoxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one ≥98%
5-Hydroxy-2-(4-hydroxyphenyl)-6-methoxy-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₂₂H₂₂O₁₁
MW: 462.4 g/mol
CAS Number: 17680-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR