5-Methylpyrimidine-2-carboxylic acid ≥98%

Supplier: Ambeed
Warning

A242921-100MG A242921-1G A242921-250MG
77748-426EA 37 USD
77748-426 77748-427 77748-428
5-Methylpyrimidine-2-carboxylic acid ≥98%
5-Methylpyrimidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MDL Number: MFCD09910349
CAS Number: 99420-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR