Methyl-5-amino-6-bromo-2-pyrazine carboxylate ≥98%

Supplier: Ambeed
Warning

A242762-1G A242762-5G
77685-682EA 176 USD
77685-682 77685-683
Methyl-5-amino-6-bromo-2-pyrazine carboxylate ≥98%
Methyl-5-amino-6-bromo-2-pyrazine carboxylate

Precaución: for Research Use Only

Formula: C₆H₆BrN₃O₂
MDL Number: MFCD09840911
CAS Number: 1076198-49-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR