Fmoc-Hyp(Bom)-OH ≥97%
Supplier: Ambeed
|
Warning
|
A310631-5G
A310631-1G
A310631-250MG
77593-864EA
132.23
USD
77593-864
77612-780
77683-918
Fmoc-Hyp(Bom)-OH ≥97%
Fmoc-Hyp(Bom)-OH
Precaución: For research use only.
Formula:
C₂₈H₃₃N₃O₈ MW: 539.6 g/mol |
MDL Number:
MFCD07366898 CAS Number: 187223-15-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...