tert-Butyl ((1R,3S)-3-aminocyclopentyl)carbamate ≥98%

Supplier: Ambeed
Warning

A310493-1G A310493-250MG A310493-5G
77571-864EA 227.63 USD
77571-864 77683-916 77683-917
tert-Butyl ((1R,3S)-3-aminocyclopentyl)carbamate ≥98%
tert-Butyl ((1R,3S)-3-aminocyclopentyl)carbamate

Precaución: For research use only.

Formula: C₁₀H₂₀N₂O₂
MW: 200.27 g/mol
MDL Number: MFCD18831392
CAS Number: 774212-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR