4-Bromo-6-fluoro-2-methyl-2,3-dihydro-1H-inden-1-one 95+%

Supplier: Ambeed
Warning

A310365-5G A310365-100MG A310365-1G A310365-250MG
77612-778EA 2408.14 USD
77612-778 77890-598 77890-599 77890-600
4-Bromo-6-fluoro-2-methyl-2,3-dihydro-1H-inden-1-one 95+%
4-Bromo-6-fluoro-2-methyl-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₁₀H₈BrFO
MDL Number: MFCD27997281
CAS Number: 892575-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR