2-(Diethylamino)ethyl 2-cyclohexyl-2-phenylacetate hydrochloride ≥98%

Supplier: Ambeed
Danger

A310094-100MG A310094-250MG A310094-50MG
77879-628EA 263 USD
77879-628 77879-629 77879-630
2-(Diethylamino)ethyl 2-cyclohexyl-2-phenylacetate hydrochloride ≥98%
2-(Diethylamino)ethyl 2-cyclohexyl-2-phenylacetate hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₃₂ClNO₂
MDL Number: MFCD00034983
CAS Number: 548-66-3
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR