4-(2-((5-Methyl-1-(naphthalen-2-yl)-1H-pyrazol-3-yl)oxy)ethyl)morpholine hydrochloride 99+%

Supplier: Ambeed
Warning

Synonyms: S1RA HCl

A242234-100MG A242234-50MG
77746-368EA 265 USD
77746-368 77746-369
4-(2-((5-Methyl-1-(naphthalen-2-yl)-1H-pyrazol-3-yl)oxy)ethyl)morpholine hydrochloride 99+%
4-(2-((5-Methyl-1-(naphthalen-2-yl)-1H-pyrazol-3-yl)oxy)ethyl)morpholine hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₄ClN₃O₂
MW: 373.88 g/mol
MDL Number: MFCD27963345
CAS Number: 1265917-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR