cis-tert-Butyl hexahydropyrrolo[3,4-b][1,4]oxazine-6(2H)-carboxylate ≥98%
Supplier: Ambeed
|
Danger
|
A309711-100MG
A309711-250MG
77883-844EA
153
USD
77883-844
77883-845
cis-tert-Butyl hexahydropyrrolo[3,4-b][1,4]oxazine-6(2H)-carboxylate ≥98%
cis-tert-Butyl hexahydropyrrolo[3,4-b][1,4]oxazine-6(2H)-carboxylate
Precaución: for Research Use Only
Formula:
C₁₁H₂₀N₂O₃ MW: 228.29 g/mol |
MDL Number:
MFCD12198674 CAS Number: 138027-02-8 UN: 3259 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...