4-Aminomethyl-7-azaindole ≥98%

Supplier: Ambeed
Warning

Synonyms: (1H-Pyrrolo[2,3-b]pyridin-4-yl)methanamine

A309367-250MG A309367-1G
77593-856EA 55.35 USD
77593-856 77672-566
4-Aminomethyl-7-azaindole ≥98%
4-Aminomethyl-7-azaindole

Precaución: for Research Use Only

Formula: C₈H₉N₃
MW: 147.177 g/mol
MDL Number: MFCD08458223
CAS Number: 888498-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR