Ethyl 5-methylpyrazolo[1,5-a]pyridine-3-carboxylate ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A242021-1G A242021-250MG
77683-731EA 86 USD
77683-731 77683-732
Ethyl 5-methylpyrazolo[1,5-a]pyridine-3-carboxylate ≥95%, Ambeed.Inc
Ethyl 5-methylpyrazolo[1,5-a]pyridine-3-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
MDL Number: MFCD06739193
CAS Number: 51135-70-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR