Irinotecan hydrochloride ≥98%

Supplier: Ambeed
Danger

A309111-100MG A309111-250MG A309111-1G
77616-346EA 61.8 USD
77616-346 77616-348 77748-601
Irinotecan hydrochloride ≥98%
Irinotecan hydrochloride

Precaución: for Research Use Only

Formula: C₃₃H₃₈N₄O₆·HCl
MDL Number: MFCD01862255
CAS Number: 100286-90-6
UN: 1544
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR