N-((9-Ethyl-9H-carbazol-3-yl)methyl)-1-methyl-1H-benzo[d]imidazol-2-amine ≥98%

Supplier: Ambeed
Warning

Synonyms: AJ2-30

A2418841-10MG A2418841-1MG A2418841-5MG
77598-350EA 237.8 USD
77598-350 77598-352 77598-354
N-((9-Ethyl-9H-carbazol-3-yl)methyl)-1-methyl-1H-benzo[d]imidazol-2-amine ≥98%
N-((9-Ethyl-9H-carbazol-3-yl)methyl)-1-methyl-1H-benzo[d]imidazol-2-amine

Precaución: for Research Use Only

Formula: C₂₃H₂₂N₄
MW: 354.45 g/mol
CAS Number: 2700322-79-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR