Ethyl 3-oxo-2-phenylpropanoate ≥95%

Supplier: Ambeed
Warning

A308862-1G A308862-5G
77879-622EA 157 USD
77879-622 77879-623
Ethyl 3-oxo-2-phenylpropanoate ≥95%
Ethyl 3-oxo-2-phenylpropanoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₃
MDL Number: MFCD11975699
CAS Number: 17838-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR