(4aS,5R,6S)-6-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-5-vinyl-4,4a,5,6-tetrahydro-1H,3H-pyrano[3,4-c]pyran-1-one ≥97%

Supplier: Ambeed
Warning

Synonyms: Sweroside

A308538-10MG A308538-1MG A308538-5MG
77601-390EA 60.48 USD
77601-390 77601-392 77601-394
(4aS,5R,6S)-6-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-5-vinyl-4,4a,5,6-tetrahydro-1H,3H-pyrano[3,4-c]pyran-1-one ≥97%
(4aS,5R,6S)-6-(((2S,3R,4S,5S,6R)-3,4,5-Trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-5-vinyl-4,4a,5,6-tetrahydro-1H,3H-pyrano[3,4-c]pyran-1-one

Precaución: for Research Use Only

Formula: C₁₆H₂₂O₉
MW: 358.34 g/mol
MDL Number: MFCD09954487
CAS Number: 14215-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR