m-PEG8-amine ≥98%

Supplier: Ambeed
Warning

A307972-1G A307972-100MG A307972-250MG A307972-25G A307972-5G
77571-856EA 52 USD
77571-856 77616-338 77616-340 77685-879 77685-880
m-PEG8-amine ≥98%
m-PEG8-amine

Precaución: for Research Use Only

Formula: C₁₇H₃₇NO₈
MDL Number: MFCD13184961
CAS Number: 869718-81-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR