GSK-J4 (Free base) 98+%

Supplier: Ambeed
Warning

Synonyms: GSK J4 hydrochloride, Ethyl 3-((6-(4,5-dihydro-1H-benzo[d]azepin-3(2H)-yl)-2-(pyridin-2-yl)pyrimidin-4-yl)amino)propanoate, N-[2-(2-Pyridinyl)-6-(1,2,4,5-tetrahydro-3H-3-benzazepin-3-yl)-4-pyrimidinyl]-β-alanine ethyl ester, GSK-J4

A307207-10MG A307207-1MG A307207-5MG
77612-768EA 51.5 USD
77612-768 77612-770 77612-772
GSK-J4 (Free base) 98+%
GSK-J4 (Free base)

Precaución: for Research Use Only

Formula: C₂₄H₂₇N₅O₂
MDL Number: MFCD22683852
CAS Number: 1373423-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR