(Chloromethyl)cyclopropane ≥95%

Supplier: Ambeed
Danger

Synonyms: Cyclopropyl methyl chloride

A307042-1G A307042-5G
77685-876EA 16 USD
77685-876 77685-877
(Chloromethyl)cyclopropane ≥95%
(Chloromethyl)cyclopropane

Precaución: for Research Use Only

Formula: C₄H₇Cl
MW: 90.55 g/mol
MDL Number: MFCD00001307
CAS Number: 5911-08-0
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR