3-Bromo-2,2-dimethyl-1-propanol ≥98%

Supplier: Ambeed
Warning

A306231-1G A306231-5G
77593-840EA 14.35 USD
77593-840 77593-842
3-Bromo-2,2-dimethyl-1-propanol ≥98%
3-Bromo-2,2-dimethyl-1-propanol

Precaución: for Research Use Only

Formula: C₅H₁₁BrO
MW: 167.05 g/mol
MDL Number: MFCD00004683
CAS Number: 40894-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR