2-(5-Bromopyrimidin-2-yl)propan-2-ol ≥97%

Supplier: Ambeed
Warning

A305431-100MG A305431-1G
77746-579EA 28 USD
77746-579 77746-580
2-(5-Bromopyrimidin-2-yl)propan-2-ol ≥97%
2-(5-Bromopyrimidin-2-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₇H₉BrN₂O
MW: 217.06 g/mol
MDL Number: MFCD19543861
CAS Number: 1193244-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR