3,5,7,8-Tetrahydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one ≥98%

Supplier: Ambeed
Danger

A305232-1MG A305232-5MG A305232-100MG A305232-10MG A305232-50MG
77601-378EA 37.92 USD
77601-378 77668-352 77879-600 77879-601 77879-602
3,5,7,8-Tetrahydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one ≥98%
3,5,7,8-Tetrahydroxy-2-(4-hydroxyphenyl)-4H-chromen-4-one

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₇
CAS Number: 527-95-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR