4-[Bis(4-iodophenyl)amino]benzaldehyde ≥96%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 4-(Bis(4-iodophenyl)amino)benzaldehyde

A305053-1G A305053-250MG A305053-5G
77683-904EA 94 USD
77683-904 77892-621 77892-622
4-[Bis(4-iodophenyl)amino]benzaldehyde ≥96%, Ambeed.Inc
4-[Bis(4-iodophenyl)amino]benzaldehyde

Precaución: For research use only.

Formula: C₁₉H₁₃I₂NO
MDL Number: MFCD20259247
CAS Number: 808758-81-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR