1-(3-Chloroanilino)-4-phenylphthalazine ≥98%

Supplier: Ambeed
Warning

Synonyms: N-(3-Chlorophenyl)-4-phenylphthalazin-1-amine, MY-5445, 1-(3-Chlorophenyl)amino-4-phenylphthalazine

A304814-10MG A304814-1MG A304814-5MG
77601-372EA 56.38 USD
77601-372 77601-374 77601-376
1-(3-Chloroanilino)-4-phenylphthalazine ≥98%
1-(3-Chloroanilino)-4-phenylphthalazine

Precaución: for Research Use Only

Formula: C₂₀H₁₄ClN₃
MW: 331.80 g/mol
MDL Number: MFCD00209838
CAS Number: 78351-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR