Methyl 4-chloro-2-formylbenzoate ≥95%

Supplier: Ambeed
Warning

A303842-1G A303842-250MG A303842-100MG
77616-316EA 87.55 USD
77616-316 77748-588 77748-587
Methyl 4-chloro-2-formylbenzoate ≥95%
Methyl 4-chloro-2-formylbenzoate

Precaución: for Research Use Only

Formula: C₉H₇ClO₃
MW: 198.6031 g/mol
MDL Number: MFCD25459256
CAS Number: 1001336-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR