Magnesium acetate ≥98%

Supplier: Ambeed
Warning

A303233-25G A303233-500G
77571-846EA 7 USD
77571-846 77593-824
Magnesium acetate ≥98%
Magnesium acetate

Precaución: for Research Use Only

Formula: (H₃CCOO)₂Mg
MDL Number: MFCD00003539
CAS Number: 142-72-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR