Dipotassium hexachloroiridate ≥98%

Supplier: Ambeed
Warning

Synonyms: Potassium hexachloroiridate(IV), Potassium hexachloroiridate (IV)

A303138-1G A303138-250MG
77601-364EA 168.1 USD
77601-364 77668-344
Dipotassium hexachloroiridate ≥98%
Dipotassium hexachloroiridate

Precaución: for Research Use Only

Formula: Cl₆IrK₂
MW: 483.13 g/mol
MDL Number: MFCD00011369
CAS Number: 16920-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR