5-((tert-Butyldimethylsilyl)oxy)pentanal ≥95%, Ambeed.Inc

Supplier: Ambeed
Danger

A302189-1G A302189-100MG A302189-250MG
77683-899EA 279 USD
77683-899 77885-802 77885-803
5-((tert-Butyldimethylsilyl)oxy)pentanal ≥95%, Ambeed.Inc
5-((tert-Butyldimethylsilyl)oxy)pentanal

Precaución: For research use only.

Formula: C₁₁H₂₄O₂Si
MDL Number: MFCD09831902
CAS Number: 87184-80-3
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR